Would you like to log in again?
Flat Nose Pliers, Aven
Supplier: AVEN TOOLS INC.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10303 10335
76245-182EA 42.7 USD InStock
76245-182 76244-516
Flat Nose Pliers, Aven
Pliers
Flat Noser pliers for bending and component handling. Box joint construction for durability and perfect jaw alignment.
  • Induction hardened jaws for durability
  • Manufactured from 400 series stainless steel
  • Dual leaf springs
  • ESD Safe cushioned grips
Dual leaf springs ensure opening and closing is comfortable. ESD Safe Grips provides protection against static as well as extra comfort for repetitive work. Mirror finished stainless steel (400 series) provides superior rust protection compared with standard carbon steel pliers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty