Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

ProcessMate™ 100 Vacuum Pickup Systems, EFD

Supplier: EFD INC.- CUSTOMER SERVICE TE
7012329
76245-224EA 2427.43 USD InStock
76245-224
ProcessMate™ 100 Vacuum Pickup Systems, EFD
Vacuum Handling Systems
The ProcessMate 100 Vacuum Pickup System provides a simple, efficient way to lift and position small or delicate components in benchtop assembly processes.

  • Comfortable, lightweight pickup pen
  • Assorted antistatic tips and vacuum cups meet a variety of applications
  • Faster, more precise placement than conventional tweezers
  • Leaves one hand free to apply adhesive, solder or other assembly fluid

It prevents damage to delicate or intricate components and saves benchtop space through its compact size.

Input AC (to power supply): Universal Multi Voltage; 64/90 VAC to 50/60 Hz
Output DC (from power supply): 24 VDC to 0.83 Amp maximum
DC Input Power Required: 8 to 20 W

Maximum Cycle Rate: 600 cycles per minute; Air Input Requirements: 7 bar (100 psi) maximum; Air Output: 0 to 110" H2O dependent on user setting.

The component can be lifted when the operator simply places the pickup pen on the component and presses an electric foot pedal to apply vacuum. When the component has been positioned, releasing the foot pedal stops the vacuum and releases the component.

Certifications: CE, RoHS, WEEE and China RoHS Compliant.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty