Would you like to log in again?
Oscilloscope Probe, Digi-Key
Supplier: Digi-Key
SP100B
76245-776EA 168.61 USD InStock
76245-776
Oscilloscope Probe, Digi-Key
Electrodes Other Electrodes
Oscilloscope Probe ensures accuracy and can improve the performance of test instrument.
  • Maximum input voltage of 600 V
  • Rise time of 3.5 ns
  • Compensation range of 10 to 35 pF
The compensation adjustment for these probes is located in the BNC to eliminate noise pickup.
Certifications: Meets IEC-1010 safety standards for 600V CAT I applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty