Would you like to log in again?
Tape 9731, Double Coated, 3M
Supplier: 3M
7010374023
76245-814CS 17400.4 USD InStock
76245-814
Tape 9731, Double Coated, 3M
Tapes
The tape is developed with a high-performance acrylic adhesive on the face side and a silicone adhesive on the liner side of a 1 mil clear PET carrier.
  • Double linered – Adhesive 350
  • Ideal for attaching silicone rubber elastomers and foam
Features acrylic and silicone adhesives for specialized applications such as silicone gasketing. A thin polyester carrier provides dimensional stability and improved handling with ease of die cutting and lamination compared to adhesive transfer tapes.
It is excellent for demanding applications where silicone materials must be bonded to a range of substrates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty