Would you like to log in again?
Envelope Gaskets, Garlock Hygienic Technologies
Supplier: Garlock
A40MPGR-200-V A40MPGR-100-V A40MPGR-150-V
76246-010EA 8.37 USD InStock
76246-010 76244-492 76246-012
Envelope Gaskets, Garlock Hygienic Technologies
Seals Gaskets
A PTFE envelope gasket has the chemical resistance of PTFE and the elastomeric characteristics of FKM Fluoroelastomer.
Certifications: Meets FDA and USP Class VI standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty