Would you like to log in again?
Pillowstat ESD Boxes, Conductive Containers
Supplier: Conductive Containers
PP1010 PP3545 PP2020 18M-1304-R1
76243-830CS 1023.9 USD InStock
76243-830 76246-070 76246-080 76243-832
Pillowstat ESD Boxes, Conductive Containers
Boxes ESD-Safe Boxes
Super soft ESD foam for surface mount technology.
  • ESD safe foam forms around the parts
  • No tooling required
  • Provides great physical protection
  • Fits multiple sized components
Ideal for fragile, sensitive and expensive components.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty