Would you like to log in again?
Mat Monitor Cord with Diode, SCS Division of Desco
Supplier: Desco Industries
2380D
76246-412EA 45.29 USD InStock
76246-412
Mat Monitor Cord with Diode, SCS Division of Desco
Cables
This cord connects workstation monitors to worksurface mats.
  • Tinned wire terminal connects to the workstation monitor
  • 10 mm stud terminal connects to the worksurface mat
  • No resistor
Features embedded diode which protects the workstation monitor for any voltage be induced onto the worksurface mat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty