Would you like to log in again?
CP STAT 100M Anti-static Bags and Films, Z-Mar Technology
Supplier: Z-Mar Technology
CPSTAT100M1012 CPSTST100MZ1518 CPSTST100M1830 CPSTST100M2929 CPSTAZ100M1012 CPSTST100M1829 CPSTST100M1216 CPSTST100M1216 CPSTST100M23Z CPSTAT100MR
76246-470CS 790.43 USD InStock
76246-470 76246-492 76246-472 76246-494 76246-510 76246-468 76246-496 76246-474 76246-386 76246-476
CP STAT 100M Anti-static Bags and Films, Z-Mar Technology
Bags Sample Bags
These bags made of a strong, abrasion resistant material offering excellent electrostatic shielding properties. The metal-in, five layer construction provides superior protection from punctures and tears.
It is free of primary amines, amides and silicone compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty