Would you like to log in again?
Anti-Static Poly Bags, Z-Mar Technology
Supplier: Z-Mar Technology
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CPSTST100M810Z CPSTST100M46Z CPSTST100M68 CPSTST100M35Z CPSTST100M58Z CPSTST100M810
76246-508CS 193.38 USD InStock
76246-508 76246-504 76246-506 76246-500 76246-502 76246-498
Anti-Static Poly Bags, Z-Mar Technology
Bags Sample Bags
Manufactured with 100% virgin low density polyethylene.
  • Contains a pink color additive
  • See through product in interior light
  • This antistatic product is amine-free
Antistatic properties meet the static decay requirements of MIL-PRF-81705E, Type III and EIA-541.
Packaging: Provided in cases.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty