Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Air Force Blow-Off Gun, Simco Ion

Supplier: Simco
Air Force™
92-6115-US
76246-748EA 3001.1 USD InStock
76246-748
Air Force Blow-Off Gun, Simco Ion
Ionizers
This is a lightweight and durable ionizingblow off gun that is used to remove targeted particle contamination.

  • Ergonomic gun design
  • Replaceable emitter point assembly and quick-eject filter
  • Strong blow-off force
  • Steady-state DC ion emission
  • IsoStat technology
  • Durable static-dissipative materials

Flexible, lightweight air hose with integral low voltage power cable and indicator light.

Air filter: 0.01 micron, Air pressure: 20 to 65 psi

Ozone level: <0.005 ppm

Certifications: CE Certified, RoHS 2 Compliant, cULus

Delivery information: Supplied with air hose, console and 120 V transformer.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty