Would you like to log in again?
Dispensing Tips, Tapered, Jensen Tools
Supplier: Jensen Tools + Supply
JG25-1.25HPTTX
76246-802PK 85.19 USD InStock
76246-802
Dispensing Tips, Tapered, Jensen Tools
Syringe Needles
Dispensing tip manufactured using the highest quality standards and the tightest possible tolerances.
  • UV-blocking additive
  • Scratch-free protection for working with sensitive parts
NT Premium tapered tips have an all-plastic tapered design to reduce backpressure and work best with medium to high viscosity materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty