Would you like to log in again?
Stencil Cleaning Roll, AGI Corporation
Supplier: AGI CORPORATION TE
176216
76246-958EA 39.18 USD InStock
76246-958
Stencil Cleaning Roll, AGI Corporation
Wipes Disposable Wipes and Towels Wipes for General Applications
This roll is a high-performance USC roll that offers excellent cleaning performance at a costeffective price.
  • High performance USC mediums
  • Developed specifically for automated under screen cleaning
  • Enhanced cleaning raises yield and throughput
  • High tear resistance raises MTBF and reduces operator intervention
  • Compatible with all DEK screen printers
The DEK SMT Roll has been developed to optimise the cleaning performance of the automated under-screen cleaning systems for DEK screen printers, and most of the MPM, EKRA, Fuji, Panasonic, Sanyo, and SMTech/Quad screen printers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty