Would you like to log in again?
Ball Mill, All-Spec Industries
Supplier: All-Spec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
115-6025
76247-048EA 26.64 USD InStock
76247-048
Ball Mill, All-Spec Industries
Mills
Carbide tipped ball mills make reaming damaged plated through holes and grinding away damaged baseboard material a snap. The 3/32" shaft fits the micro drill system hand piece and the mill handle.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty