Would you like to log in again?
Crescent® Diagonal Compound Action Plier, Apex
Supplier: Apex
Crescent®
PS5429C
76247-242EA 67.82 USD InStock
76247-242
Crescent® Diagonal Compound Action Plier, Apex
Pliers
This plier has compound leverage design that increases cutting power by 50%.
  • Lighter than standard pliers, reduces fatigue
  • Co-molded grips for added comfort and superior control
  • Self – opening
  • Induction hardened edges for long cutting life
Made of chrome vanadium steel for increased durability. Features with heat treated joint rivet for smooth and consistent operation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty