Would you like to log in again?
ESD Transport Carts, Bomir
Supplier: BOMIR INC TE
ATC-1 ATC-1S
76247-800EA 1931.88 USD InStock
76247-800 76247-342
ESD Transport Carts, Bomir
Carts
Platforms are made of conductive reinforced ABS material and wheels are made of ESD-conductive material.
  • ESD safe stainless steel construction
  • Six ESD safe wheels for easy stairs climbing
  • Loading capacity of 150 kg and for top shelf 50 kg
The cart has a 10 mm snap for grounding the cart at the parking station on the upper part of the right/front leg. Six feet grounding coil cord terminated with banana plug and an alligator clip is included with the cart.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty