Would you like to log in again?
D-Wipe® Towel, ESCA Tech
Supplier: ESCA TECH INC. TE
WT150
76247-404CS 341.75 USD InStock
76247-404
D-Wipe® Towel, ESCA Tech
Wipes Disposable Wipes and Towels Paper Towels
D-Wipe® disposable towels are generously saturated with a gentle pH balanced cleaner to quickly remove lead and heavy metal dusts.
The soft towels lift, bind and hold lead and other heavy metal dusts and take them away without smearing. With Aloe skin moisturizers, D-Wipe towels are the portable and convenient way to clean the hands, arms, face and body.
Packaging: Available in canisters or individually wrapped single towels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty