Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sterile 70% Isopropyl Alcohol with 30% USP Grade DI Water for Cleanroom Cleaning, FG Clean Wipes

Supplier: FG Cleanwipes

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

6LS7030VSIPA32B
76247-550CS 824.13 USD InStock
76247-550
Sterile 70% Isopropyl Alcohol with 30% USP Grade DI Water for Cleanroom Cleaning, FG Clean Wipes
Cleaning Agents Disinfectants
70% USP Grade Isopropyl Alcohol / 30% USP Grade Deionized Water, Filtered to 0.2 um.

  • 32 oz non-atomizing spray bottle for controlled dispensing to surfaces
  • Eliminates the need for mixing and processing sterile alcohol solution "in-house"
  • Gamma irraiated and validated to a sterility level of 10-6
  • Pyrogen-free
  • Each bottle is double-bagged for cleanroom use

Recommended for use in ISO 3-8 controlled environments.

Certifications: Certificate of Sterility Included.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty