Would you like to log in again?
Scorpion3 High Performance Ionizing Bar with 19 Tungsten Emitters, Simco Ion
Supplier: Simco Ion
4011550
76247-602EA 5785.33 USD InStock
76247-602
Scorpion3 High Performance Ionizing Bar with 19 Tungsten Emitters, Simco Ion
Ionizers
This is a 19 emitters Scorpion3™ 24 V dc rated voltage tungsten ion bar. It is designed for use in a wide variety of critical environments that require static and contamination control including semiconductor and medical device areas.
  • Emitter pin available
  • LED and remote alarm indication
  • IR remote for easy calibration
  • Daisy chain capability
Certifications: CE Certified, cULus
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty