Would you like to log in again?
Furon® UPX 2000 Pneumatically Actuated Diaphragm Valve, Ryan Herco Flow Solutions
Supplier: Ryan Herco Flow Solutions
UPX2-F88NC
76247-782EA 2381.1 USD InStock
76247-782
Furon® UPX 2000 Pneumatically Actuated Diaphragm Valve, Ryan Herco Flow Solutions
Valves Vacuum Valves
This type of diaphragm valve provides superior life expectancy over competitive valves. Fully swept internal flow path with no entrapment areas to eliminate bacteria growth.
  • All virgin PTFE/high purity molded PFA wetted flow path
  • PTFE spring isolation bellows
  • No exposed metallics
  • Visual position indication
  • Assembled and double bagged in class 100 clean room
Feature with double diaphragm containment with leak detection capability. Utilizes the Furon® SCR spring to assure reliability in the harshest HCI applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty