Would you like to log in again?
SMD Parallel Remover Handpiece, American Hakko Products
Supplier: American Hakko Products
FM2022-02
76248-238EA 712.16 USD InStock
76248-238
SMD Parallel Remover Handpiece, American Hakko Products
Soldering Supplies Soldering Irons
Parallel Remover uses revolutionary parallel action between the tip ends-one tip remains stationary allowing greater surface contact between the tip and the component and providing more efficient and even heat transfer.
  • Parallel Remover uses the same quick-change composite tip technology
  • Fixing collar properly aligns the tips opposite each other
  • Changing tips for different types of components
Removing small chips or larger SOP components, the parallel action of the FM-2022 also allows for a more secure hold onto the component.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty