Would you like to log in again?
Pneumatic Desoldering Module, JBC
Supplier: JBC
MVE-A
76248-306EA 2232.32 USD InStock
76248-306
Pneumatic Desoldering Module, JBC
Soldering Supplies Desoldering Tools
The pneumatic desoldering module is indispensable for a complete desoldering process.
  • Clean and dry compressed air is required
  • ESD Safe
  • Operating temperature: 10 to 40 °C
The usage of compressed air and a Venturi valve, the system provides the best results due to its instantaneous vacuum ramp.
This module can be joined to any port of the DDE and DME control units and has the possibility to be controlled by the pedal connection.
Delivery information: Supplied with easy-to-change filters for extending the pumps life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty