Would you like to log in again?
Standard Hand Tap, Sid Tool
Supplier: Sid Tool
68762897
76249-004EA 1093.76 USD InStock
76249-004
Standard Hand Tap, Sid Tool
Tool Sets
Straight four flute standard hand tap, with bottoming chamfers are used for threading blind holes. They have 1 to 2 threads.
  • High speed steel (HSS) provides good wear-resistance
  • Used in general purpose applications for both ferrous and nonferrous materials
  • Bright provides a smooth, polished finish on the tool
It increases chip flow in softer materials such as aluminum, wood and plastic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty