Would you like to log in again?
Morse® Cobalt Aircraft Type J Jobber Length Drills, Cutting Tool Supply
Supplier: Cutting Tool Supply
17112-MO
76249-146EA 6.93 USD InStock
76249-146
Morse® Cobalt Aircraft Type J Jobber Length Drills, Cutting Tool Supply
Tool Sets
Heavy duty construction. Cobalt steel offers increased hardness, toughness, wear resistance and heat resistance.
  • 135° split point
Recommended for drilling tough, high tensile strength materials that generate higher cutting temperatures including high alloy steels, ferrous castings, titanium, inconel, stainless steels and other difficult-to-drill material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty