Would you like to log in again?
Stopper Assembly, Newage Industries
Supplier: NewAge Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CA7311462 CA7311463
76249-278EA 325.25 USD InStock
76249-278 76249-276
Stopper Assembly, Newage Industries
Stoppers Silicone Stoppers
This is a custom stopper assembly molded, GL45 with two holes.
Used for silicone pump tubing.
Packaging: Double bagged with label on inner bag.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty