Would you like to log in again?
Weller® WX Soldering Station, Apex
Supplier: Apex
Weller®
WX2021N
76249-356EA 4251.54 USD InStock
76249-356
Weller® WX Soldering Station, Apex
Soldering Supplies Soldering Stations
This WX soldering station with an elegant design and solid improvements in performance offers the perfect combination of features to help the operation become more efficient and productive.
  • Very high capacity
  • Quick heating-up time
  • Capacitive touchscreen made of real glass
  • Antistatic, chemical and temperature resistant
  • Graphic LC display to view text and graphic elements from all angles
  • Blue LED background lighting for improved legibility
Standard digital connection and decoupled robot connection manages fume extraction units, heating plates, programmable logic controller (PLC). Pencil has built-in analog-to-digital converter and data memory read and re-calibrated pencil eliminates need to calibrate soldering station.
The Spring system provides steady contact pressure with antistatic soft grip. Tips can be changed without tools.
Automatic standby mode activated by a sensor in handle that detects when device is not in use. No special stop-and-go rest required. Automatic standby mode sensor also controls all connected accessories such as fume extractors and heating plates.
Delivery information: Includes soldering station with micro soldering iron, 40 W/12 V safety rest WDH 50 and micro tweezer, 2× 40 W/12 V safety rest WDH 60.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty