Would you like to log in again?
ESD Safe Dial Caliper DialMax, Wiha Tools
Supplier: Wiha Tools
41105
76249-448EA 325.18 USD InStock
76249-448
ESD Safe Dial Caliper DialMax, Wiha Tools
Calipers
ESD Safe Dial Caliper DialMax - metric and inch non-metallic high-tech material with 60% fiber glass content.
  • For working in sensitive electronic components
  • Conductive, anti-static, dissipative
For outside, inside, depth and step measurements. Impact resistant dial can be recalibrated to zero.
Certifications: ESD Standard IEC 61340-5-1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty