Would you like to log in again?
Memphis Dyneema® Cut Resistant Gloves, MCR Safety, Zoro
Supplier: MCR Safety
Memphis Dyneema®
9677-S
76249-550PR 46.21 USD InStock
76249-550
Memphis Dyneema® Cut Resistant Gloves, MCR Safety, Zoro
Gloves
Ideal for automotive, metal fabrication, ceramics, paper and pulp, textile industries and other applications requiring ultimate cut protection.
  • Gunn cut
  • Cut and abrasion resistant
  • Full finger style
Polyurethane has been used as coating material.
Certifications: Meets ANSI/ISEA Cut Level A3 and ANSI/ISEA Abrasion Level 5.
ANSI 105, EN 388 certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty