Would you like to log in again?
Static Shielding Bags, 81705 Series, SCS, Desco Industries
Supplier: Desco Industries
817810
76249-722PK 30.73 USD InStock
76249-722
Static Shielding Bags, 81705 Series, SCS, Desco Industries
Bags Storage Bags
These static shield bags are manufactured from four layers - static dissipative coating, polyester, metal, and polyethylene laminate.
  • Polyester dielectric in concert with the metal layer provides discharge shielding
  • Exterior is static dissipative and allows electrostatic charges to be removed when grounded
Film printing is a buried layer that will not rub-off or become removed with chemicals like IPA.
Certifications: MIL-PRF-81705E Type III, Class 2. Lead-free RoHS 2, REACH and Conflict Minerals Compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty