Would you like to log in again?
End Mill Adapters, Cutting Tool Supply
Supplier: Cutting Tool Supply
CAT40-EM025-0250 CAT40-EM018-0250 CAT40-EM012-0250
76249-904EA 314.02 USD InStock
76249-904 76249-902 76249-900
End Mill Adapters, Cutting Tool Supply
Adapters Adapters, Conical Ground Joints
End mill adapters features H5 bore tolerance, coolant thru center and DIN AD+B coolant.
  • Features bore runout of 3 µm - 0.0001"
  • Balanced G6.3 at 20000 rpm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty