Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cleanroom Stool, ISO 4

Supplier: BevCo Precision Manufacturing
Versa
3210C2-V-BLK
76249-954EA 852.12 USD InStock
76249-954
Cleanroom Stool, ISO 4
Chairs
Versa ISO 5 black vinyl tubular steel stool.

  • 16" Ø vinyl seat that complies with CAL 117
  • The pneumatic height adjustment allows for stress-free seat height changes from 19 - 24"
  • The stool features a sturdy 22" Ø chrome tubular steel base with welded footring
  • Model comes standard with mushroom glides
  • 300 lb. weight capacity

The sturdy construction and modern design of the Bevco Versa ISO 5 cleanroom backless stool is ideal for a wide range of applications, including medical, laboratory, etc.

ANSI/BIFMA; GreenGuard Gold.

Ordering information: Optional casters can be ordered separately. 12 - year warranty.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty