Would you like to log in again?
Sp Suction lens 6D, Vision Engineering
Supplier: Vision Engineering
SPD025979
76250-230EA 218.97 USD InStock
76250-230
Sp Suction lens 6D, Vision Engineering
Magnifiers
STAYS (Self-Traction Accessory Yield System) Lenses are for use with KFM LED, KFM, WAVE LED, WAVE+Plus, LFM LED and Circus Magnifiers.
Simply moisten the silicon gasket around the lens and gently press down to affix the STAYS lens to the magnifier primary lens. The lens itself has a 2" diameter (2.5" including the silicon gasket frame).
6-Diopter STAYS Lens: When used with a 3D/3.5D lens the working distance is 4". When used with a 5D lens the working distance is 3.8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty