Would you like to log in again?
Single Rod Hooks, Triton Products
Supplier: Triton Products
51013 51119
76250-320PK 8.45 USD InStock
76250-320 76250-318
Single Rod Hooks, Triton Products
Furniture Shelves
Single rod hooks are offered with multiple degree bends to accommodate any size tool assuring items will not slide off.
  • Hooks mount vertically and horizontally in four distinct positions
  • Two and four-point contact locking mechanism offers industrial strength holding capability
  • Fixtures install from the front, so no need to remove the board for installation or repositioning of LocHooks
  • Compatible with all LocBoard sizes and other brands of steel tool boards with square holes spaced 11/2" O.C.
Hang wrenches, putty knives, paint brushes, carpenters level, gardening tools and even the kitchen pans (any item having an open-hole handle).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty