Would you like to log in again?
Heat Resistant Gloves, Nitrile, Zoro
Supplier: Zoro
G0055772
76250-624PR 92.09 USD InStock
76250-624
Heat Resistant Gloves, Nitrile, Zoro
Gloves
These heat resistant gloves are function specific.
  • Gun cut
  • Wing thumb style
Applications include bottling and glass handling, handling large panes of hot glass, intermittent handling of hot cores, castings, fabricated metals and controls up to 400 °F.
Certifications: EN 407.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty