Would you like to log in again?
MCR Safety Cut Resistant Glove, Model 9389, Zoro
Supplier: Zoro
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9389S
76250-880PR 53.65 USD InStock
76250-880
MCR Safety Cut Resistant Glove, Model 9389, Zoro
Gloves
Ideal for aerospace, construction, metal, plastics, recycling, automotive, glass, pulp and paper, public utilities and salvage industrial applications.
  • Full finger style
  • Gunn cut
  • Straight thumb
Glove is coated with natural rubber latex.
Certifications: Meets ANSI/ISEA Cut Level A4 and ANSI/ISEA Abrasion Level 3. EN 388, EN 511, CE certified.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty