Would you like to log in again?
DIP Liner, American Hakko Products
Supplier: American Hakko Products
FT100
76250-890EA 118.96 USD InStock
76250-890
DIP Liner, American Hakko Products
Soldering Supplies Desoldering Tools
Dip liner allows the operator to simply insert the IC case into the Dipliner chute, and the leads are protected right through circuit board insertion.
  • Low cost and high performance
  • No more broken leads during IC and LSI insertion or transportation
Adjustable to fit all types, sizes, and makes of ICs and LSIs and possess with simple mechanism for maintenance-free operation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty