Would you like to log in again?
CHP Micro Nipper, American Hakko Products
Supplier: American Hakko Products
TR-20-SM
76251-002EA 35.75 USD InStock
76251-002
CHP Micro Nipper, American Hakko Products
Cutters
These precision cutters are sheared from 2.0 to 5.0 mm carbon steel, then precision ground to exact specifications.
  • 2.0 mm steel
  • Flush cut
  • 20 AWG max cut (determined with copper wire)
  • Rounded outside edges to reduce nicking on adjacent components and leads
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty