Would you like to log in again?
Indicator Labels, Drop-N-Tell, Uline
Supplier: Uline
S-867
76251-162PK 124.2 USD InStock
76251-162
Indicator Labels, Drop-N-Tell, Uline
Labels
Indicates shock damage at a glance.
  • Non-resettable
Attach self-stick detector to product or package exterior. 25G range for most electronic equipment, glassware. Included warning labels affix to package and bill of lading.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty