Would you like to log in again?
Ultrabake™ Silicone Caps, SC-SH Series, Caplugs
Supplier: Caplugs
SC125-0500 SC223-0500 SC37-0500 SC93-0500 SC218 SC109-0500 SC1125 SC295-0500 SC223-1000 SC020-0500 SC172 SC93 SC172-0500 SC970 SC125 SC156-0500 SC148 SC456-1000 SC437 SC70 SC295
76250-542PK 394.93 USD InStock
76250-542 76247-668 76250-552 76250-540 76252-652 76248-780 76248-782 76246-964 76248-860 76248-766 76248-886 76248-768 76248-548 76248-858 76251-750 76247-200 76251-526 76247-462 76247-670 76247-688 76251-748
Ultrabake™ Silicone Caps, SC-SH Series, Caplugs
Fittings and Connectors
SC-SH Series Ultrabake™ Silicone Caps are designed to mask plain and threaded studs, tube ends and even slotted holes; they will mask off the stud and allow finishing up to it. Recommended for powder coating, e-coating, plating and anodizing.
  • Recommended temperatures up to 600 °F (315 °C)
  • Rugged and durable
  • Can stretch to mask larger dimensions
  • Reusable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty