Would you like to log in again?
Burpless® Steel Drum Funnel, PIG®
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DRM1125-RD-NPT
76251-828EA 947.04 USD InStock
76251-828
Burpless® Steel Drum Funnel, PIG®
Funnels Filtration Funnels Drum Funnels
Steel drum funnel with Multiple lid color (Red, Black, Yelow) options let you segregate drums, lockable to prevent unauthorized access.
  • Fast pouring, no splashback and a one-hand latching lid make it easy
  • Exclusive latching handle design easily opens and closes lid in a simple, single-handed operation
  • Secure closure helps you meet closed container regs, but opens and closes instantly
Can pour without venting through 3/4" bung, a possible closed container reg violation if left open.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty