Would you like to log in again?
Pin Gages, Plus, Class ZZ, Zoro
Supplier: Zoro
111118000 111124000 111122700
76252-086EA 22.11 USD InStock
76252-086 76252-082 76252-084
Pin Gages, Plus, Class ZZ, Zoro
Measuring and Leveling Tools
Pin Gage, Plus, Class ZZ.
  • Tolerence of +0.0002"
  • Steel finish
Made of 52100 tool steel 60/65 Rc.
Delivery information: Includes Certificate of Accuracy Traceable to NIST.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty