Would you like to log in again?
Lista™ Xpress Mobile Cabinet, Arlink
Supplier: Arlink
Lista™ Xpress
XSHS0900-0903M XSST0750-0602M XSMP0750-0601M
76251-478EA 0.73 USD InStock
76251-478 76251-480 76252-328
Lista™ Xpress Mobile Cabinet, Arlink
Furniture Storage Cabinets
Lista Xpress mobile cabinet provides safe and secure storage and transport for parts and tools.
  • PrevenTip® locking system – one-drawer-at-a-time feature prevents accidental tipping
  • Each drawer includes individual drawer latch to secure it in closed position for safety when moving
  • Create keyed-alike cabinet groupings
  • Available from individual removable core locks, keyed-alike and master key systems
Delivery information: Unit includes mobile base with two fixed and two swivel 6×2" casters with wheel locks, and handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty