Would you like to log in again?
Pink Cushion Grade Dissipative Foam, Protektive Pak®
Supplier: Desco Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
37713
76252-560EA 282.34 USD InStock
76252-560
Pink Cushion Grade Dissipative Foam, Protektive Pak®
Wipes Sponges
Static dissipative polyurethane pink foam is antistatic and low charging. It minimizes electrostatic charge generation shock.
  • Static dissipative polyurethane: <1×10¹¹ ohms
  • Not recommended for lead insertion applications
  • Easy to adapt for custom use by die-cutting, laminating and more
  • Non-contaminating, non-corrosive, and non-sloughing
  • Density: 1.4 lbs./ft.
Antistatic, low charging foam minimizes electrostatic charge generation shock. Not recommended for lead insertion applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty