Would you like to log in again?
Orifice Plate Gasket, Garlock Hygienic Technologies
Supplier: Garlock
A80MPSFY-150-GAS
76252-636EA 52.06 USD InStock
76252-636
Orifice Plate Gasket, Garlock Hygienic Technologies
Seals Gaskets
Orifice plates can advance system’s performance, adjust flow rates, balance backflow and equalize backpressure during SIP procedures. Achievable benefits while maintaining sanitary conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty