Would you like to log in again?
Statfree DPL Plus™ Diamond Plate Floor Mats, Desco
Supplier: Desco Industries
40981 40979
76252-808EA 516.04 USD InStock
76252-808 76246-454
Statfree DPL Plus™ Diamond Plate Floor Mats, Desco
Floor Mats Permanent Floor Mats
These floor mats provide better traction especially where moisture may accumulate, with highly visible yellow border with beveled edges and radius corners.
  • Consistent electrical and slip resistant
  • Foam rubber base will not break down or absorb moisture
  • Allows carts and chairs to roll over without damage
It maintains consist path to ground when on operator is on properly grounded conductive floor mat.
Certifications: Exceeds ANSI/ESD S20.20
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty