Would you like to log in again?
Sticklers® CleanClicker™ End-Face Cleaners, Microcare
Supplier: MicroCare
MCC-CCU125 MCC-CCU250 MCC-CCR125
76247-912EA 427.25 USD InStock
76247-912 76247-914 76252-936
Sticklers® CleanClicker™ End-Face Cleaners, Microcare
Wipes Cleaning Kits and Pads
The Sticklers® CleanClicker™ 1.25 mm and 2.5 mm fiber optic connector cleaners are the latest cleaning innovation from the experts at Sticklers. These rugged, precision cleaners use a smooth click-to-clean push-action with light tip forces to reduce the risk of end-face damage.
  • Easy-to-use on outside plant and premise networks
  • Part of the industry’s most complete line of fiber optic cleaning products
  • Color coded labels and packaging minimize user error and speed cleaning
  • Cleans dust, particulate and oil contamination
With over 750 cleans, the Sticklers CleanClicker is perfect for cleaning any size network, any installation, from single to thousands of connectors.
Sticklers CleanClicker cleaners are packaged color-coded byconnector size to help users quickly ensure they have the right cleaner for the connector at hand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty