Would you like to log in again?
LC NSF Series Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
71400 72200 74900 71200 71200
76251-620EA 604.21 USD InStock
76251-620 76252-952 76251-644 76251-622 76251-618
LC NSF Series Couplings, Colder Products Company
Fittings and Connectors
LC Series chrome-plated brass couplings are built tough and made to last in the most demanding applications.
  • Operating temperature: −40 to 82 °C
  • Valve spring is made of 316 stainless steel
  • Can withstand in high temperature and pressure
These couplings offer the flexibility of multiple configurations and terminations to mate with both the PLC acetal and PLC12 polypropylene couplings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty