Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KD® High Precision Static Dissipative Tweezers, TDI International

Supplier: TDI International
2AKDR-SA
76253-086EA 233.72 USD InStock
76253-086
KD® High Precision Static Dissipative Tweezers, TDI International
Forceps
High precision ceramic tip tweezers provide enhanced strength with high surface finish ceramic tip material and anti-magnetic stainless steel handles. These tweezers are excellent for ultra high temperature applications and handling of sensitive ceramic and glass parts where nonmetallic scratching is critical.

  • Surface resistivity 10⁶ to 10⁸ Ohms/sq. at 50V and 100V threshold
  • Surface resistivity 10⁷ to 10⁹ Ohm/sq. at 10V threshold
  • Extreme high-temperature tip resistance: 1000 °C
  • Solder, chemical, solvent & acid resistant
  • Lead free

Ergonomic ESD safe static dissipative cushion grips reduce stress and tension on fingers, while providing superior operator comfort when working continuously with small parts. Static dissipative zirconia ceramic tips – 'KD'® with excellent transient current discharge performance.

Alignable replacement tips and enhanced break strength and surface finish. High precision fine tip symmetry and balance with stainless anti-magnetic handles.

Certifications: Class 0 CDM Compliant.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty