Would you like to log in again?
Compact Router Kit, Global Industrial
Supplier: GLOBAL INDUSTRIAL TE
RT0701C
76253-172EA 478.28 USD InStock
76253-172
Compact Router Kit, Global Industrial
Tool Sets
The 11/4 HP Compact Router Kit delivers performance, versatility and precision in a compact size.
  • Soft start feature for smooth start ups
  • Accepts industry standard template guides
  • Electronic speed control maintains constant speed under load
  • Heavy duty aluminum motor housing engineered for increased durability
Variable speed control dial (10000 - 30000 RPM) to match the speed to the application. Base can be removed quickly to install other bases (plunge, tilt and offset).
Quick release cam lock system for convenient depth adjustments and base removal/installation. Smooth rack-and-pinion fine depth adjustment system for more precise settings.
With a full maximum horsepower and three bases included in the kit, it is engineered for a full range of precision routing applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty