Would you like to log in again?
Cutters, Cut and Bend Leads, Excelta
Supplier: Excelta
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
907-7 530DE-US 500-108-US 500-108-US 907 530FE-US
63042-404EA 415.21 USD InStock
63042-404 63042-548 63042-528 63043-038 63043-016 76253-250
Cutters, Cut and Bend Leads, Excelta
Pliers
Designed to cut leads on PC board and bend them to keep them in place.
  • Anti-shock trimming of leads to specific length
  • Bends the lead to keep placement on pc board
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These shear cutting and bending plier are designed to cut the leads to a specific size and then bend or swage them to keep the component in place when soldering. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. antimicrobial static dissipative (10⁸ ohms/sq) ergonomic grips with dual leaf springs. These tools are made of carbon tool steel with flame hardened cutting edges.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty