Would you like to log in again?
Gearwrench® Deep SAE Socket, 6 Point, Apex
Supplier: Apex
80370D
76253-320EA 16.24 USD InStock
76253-320
Gearwrench® Deep SAE Socket, 6 Point, Apex
Tool Sets
This is 3/8" Drive 6 point deep SAE socket with off-corner loading design reduces fastener rounding.
  • Chamfered opening to guide fastener into socket
  • Large hard-stamped size identification
  • Double Line (SAE) to easily identify standard vs. metric sockets
Full polish chrome finish cleans easily and resists corrosion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty